This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNE2 SIRNA Assays ›
Gene Symbol:
Kcne2
Gene Name:
potassium channel, voltage-gated Isk-related subfamily E regulatory beta subunit 2
Gene Aliases:
Mirp1
Chromosome Location:
Chr.11: 32434786 - 32447264 on Build Rnor_6.0
UniGene ID:
Rn.48691
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS350880
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcne2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133603.2 NP_598287.1 2 480
XM_006248023.2 XP_006248085.1 2 906
XM_008768554.1 XP_008766776.1 2 716
GenBank mRNA AF071003.1 - 2 321
BC089778.1 - 2 480 7301610

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

aging
potassium ion import
cellular protein localization
cellular response to drug
tongue development
regulation of membrane repolarization
regulation of ventricular cardiac muscle cell membrane repolarization
potassium ion export
potassium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
ventricular cardiac muscle cell action potential
membrane repolarization
membrane repolarization during action potential
regulation of heart rate by cardiac conduction
regulation of potassium ion transmembrane transport
positive regulation of voltage-gated calcium channel activity
positive regulation of proteasomal protein catabolic process
regulation of inward rectifier potassium channel activity
regulation of cyclic nucleotide-gated ion channel activity
regulation of delayed rectifier potassium channel activity
negative regulation of voltage-gated potassium channel activity
inward rectifier potassium channel activity
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
cation channel activity
protein binding
potassium channel regulator activity
protein homodimerization activity
ion channel binding
voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm