This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other INPP5J SIRNA Assays ›
Gene Symbol:
Inpp5j
Gene Name:
inositol polyphosphate-5-phosphatase J
Gene Aliases:
Inpp, Pib5pa, Pipp
Chromosome Location:
Chr.14: 83730861 - 83742011 on Build Rnor_6.0
UniGene ID:
Rn.23872
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS350825
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Inpp5j
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133562.1 NP_598246.1 2 509
XM_006251155.2 XP_006251217.1 2 530
XM_008770248.1 XP_008768470.1 2 551
GenBank mRNA AB032551.1 - 2 509

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of neuron projection development
dephosphorylation
negative regulation of microtubule polymerization
negative regulation of peptidyl-serine phosphorylation
phosphatidylinositol dephosphorylation
inositol-polyphosphate 5-phosphatase activity
SH3 domain binding
phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
inositol-1,4,5-trisphosphate 5-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0