This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMLHE SIRNA Assays ›
Gene Symbol:
Tmlhe
Gene Name:
trimethyllysine hydroxylase, epsilon
Gene Aliases:
Tmlh
Chromosome Location:
Chr.20: 237461 - 285125 on Build Rnor_6.0
UniGene ID:
Rn.3607
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS350748
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmlhe
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133387.2 NP_596878.2 6, 7 871
XM_006255855.1 XP_006255917.1 5, 6 880
GenBank mRNA AF374406.1 - 4, 5 712
BC085923.1 - 6, 7 871 7109209

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_138758, AY033513, AK052425, BC010495, AK144627, and others.

Gene Ontology Categories:

Function(s) Process(es)

carnitine biosynthetic process
negative regulation of oxidoreductase activity
oxidation-reduction process
iron ion binding
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
L-ascorbic acid binding
trimethyllysine dioxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0