This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DYNLRB1 SIRNA Assays ›
Gene Symbol:
Dynlrb1
Gene Name:
dynein light chain roadblock-type 1
Gene Aliases:
Dncl2a, km23
Chromosome Location:
Chr.3: 150761329 - 150782493 on Build Rnor_6.0
UniGene ID:
Rn.3560
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS350734
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dynlrb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_131910.3 NP_571985.1 4 392
GenBank mRNA AF073839.2 - 4 315
AY026512.1 - 4 392
BC058437.1 - 4 333 6921076

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

visual behavior
retrograde axonal transport
metabolic process
motor activity
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0