siRNA Details
Gene Symbol: Rcvrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080901.1 | NP_543177.1 | 1 | 56 | ||
GenBank mRNA | AY063482.1 | - | 1 | 56 |

Gene Symbol: Rcvrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080901.1 | NP_543177.1 | 1 | 56 | ||
GenBank mRNA | AY063482.1 | - | 1 | 56 |