This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other H3F3B SIRNA Assays ›
Gene Symbol:
H3f3b
Gene Name:
H3 histone, family 3B
Gene Aliases:
H3.3B
Chromosome Location:
Chr.10: 104573666 - 104575890 on Build Rnor_6.0
UniGene ID:
Rn.124815
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS350467
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: H3f3b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053985.2 NP_446437.1 3 396
GenBank mRNA BC063159.1 - 3 396 6921452
BC078759.1 - 3 365 7112262
FQ215473.1 - 3 389
FQ225133.1 - 3 391
FQ225528.1 - 3 389
FQ226787.1 - 3 389
FQ227040.1 - 3 391
FQ227260.1 - 3 394
FQ227957.1 - 3 391
FQ228569.1 - 3 391
FQ232451.1 - 3 325
FQ232671.1 - 3 389
FQ232852.1 - 3 391
FQ234172.1 - 3 391
FQ234408.1 - 3 389
FQ235236.1 - 3 394
X73683.1 - 3 387

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008211, AK168698, AK044933, AK169226, AK159807, and others.

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
nucleosome assembly
DNA replication-independent nucleosome assembly
nucleus organization
brain development
response to hormone
positive regulation of cell growth
muscle cell differentiation
regulation of centromere complex assembly
negative regulation of chromosome condensation
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
molecular_function
DNA binding
nucleosomal DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0