This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HMGN3 SIRNA Assays ›
Gene Symbol:
Hmgn3
Gene Name:
high mobility group nucleosomal binding domain 3
Gene Aliases:
-
Chromosome Location:
Chr.8: 90628203 - 90665059 on Build Rnor_6.0
UniGene ID:
Rn.106652
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS350130
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hmgn3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007020.2 NP_001007021.1 3, 4 218
NM_001201353.1 NP_001188282.1 3, 4 218
NM_001201354.1 NP_001188283.1 3, 4 218
NM_001201355.1 NP_001188284.1 3, 4 218
XM_006243445.2 XP_006243507.1 3, 4 723
XM_006243446.2 XP_006243508.1 3, 4 723
XM_008766200.1 XP_008764422.1 3, 4 723
GenBank mRNA BC082036.1 - 3, 4 173 7130894
CB751211.1 - 3, 4 101
CB774488.1 - 3, 4 186
FM058030.1 - 3, 4 218

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
chromatin modification
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of insulin secretion involved in cellular response to glucose stimulus
chromatin binding
nucleosomal DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0