This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NOL12 SIRNA Assays ›
Gene Symbol:
Nol12
Gene Name:
nucleolar protein 12
Gene Aliases:
Nop25, RGD1305327
Chromosome Location:
Chr.7: 120161238 - 120166852 on Build Rnor_6.0
UniGene ID:
Rn.17454
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS342765
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nol12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012747.2 NP_001012765.1 2 149
XM_006242007.2 XP_006242069.1 2 425
GenBank mRNA AY917132.1 - 2 127
BC103643.1 - 2 149 7301307
FQ227943.1 - 2 197

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133800, AK083539, BC094424, AK167446, AY940114, and others.

Gene Ontology Categories:

Process(es)

rRNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0