This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRR3 SIRNA Assays ›
Gene Symbol:
Prr3
Gene Name:
proline rich 3
Gene Aliases:
Cat56
Chromosome Location:
Chr.20: 3300190 - 3307262 on Build Rnor_6.0
UniGene ID:
Rn.94992
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS342615
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prr3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212544.2 NP_997709.1 4 544
XM_006255942.1 XP_006256004.1 5 612
XM_006255943.2 XP_006256005.1 4 430
GenBank mRNA BC098636.1 - 5 622
BC126073.1 - 4 544 7311139

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_145487, NM_001282012, NM_001282024, NM_001282013, AJ504719, and others.

Gene Ontology Categories:

Process(es)

poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0