This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ERCC8 SIRNA Assays ›
Gene Symbol:
Ercc8
Gene Name:
excision repair cross-complementation group 8
Gene Aliases:
Ckn1
Chromosome Location:
Chr.2: 39434617 - 39473392 on Build Rnor_6.0
UniGene ID:
Rn.107979
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS342160
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ercc8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107650.2 NP_001101120.2 6, 7 599
XM_008760728.1 XP_008758950.1 5, 6 509
GenBank mRNA BC161924.1 - 5, 6 474 8370925

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_000082, AK304610, AK294856, AK056931, AK226129, and others.
This siRNA also targets the following transcripts in Mouse: NM_028042, BC056463, AK157504, AK157175, AK014517, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
biological_process
response to UV
response to X-ray
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA repair
protein autoubiquitination
molecular_function
DNA helicase activity
ubiquitin-protein transferase activity
DNA-dependent ATPase activity
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0