This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C1QTNF1 SIRNA Assays ›
Gene Symbol:
C1qtnf1
Gene Name:
C1q and tumor necrosis factor related protein 1
Gene Aliases:
Ctrp1
Chromosome Location:
Chr.10: 107455845 - 107477160 on Build Rnor_6.0
UniGene ID:
Rn.53880
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS341963
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1qtnf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007675.1 NP_001007676.1 4 814
XM_006247800.2 XP_006247862.1 4 1340
XM_006247801.2 XP_006247863.1 4 955
XM_006247802.2 XP_006247864.1 4 1412
XM_008768374.1 XP_008766596.1 4 1016
GenBank mRNA BC083720.1 - 4 814 7190841
FQ209955.1 - 4 915

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytosolic calcium ion concentration
negative regulation of platelet activation
positive regulation of gene expression
regulation of glucose metabolic process
positive regulation of MAPK cascade
protein homooligomerization
positive regulation of protein kinase B signaling
protein heterotrimerization
negative regulation of platelet aggregation
positive regulation of aldosterone secretion
collagen binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0