This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC3 SIRNA Assays ›
Gene Symbol:
Zdhhc3
Gene Name:
zinc finger, DHHC-type containing 3
Gene Aliases:
-
Chromosome Location:
Chr.8: 132158418 - 132204542 on Build Rnor_6.0
UniGene ID:
Rn.8573
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS341853
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zdhhc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039014.1 NP_001034103.1 1 56
XM_006244188.2 XP_006244250.1 2 229
XM_008766714.1 XP_008764936.1 2 233
XM_008766715.1 XP_008764937.1 2 618
XM_008766716.1 XP_008764938.1 3 444
GenBank mRNA AY886522.1 - 1 56
BC128697.1 - 2 175 7444822

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0