This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP36L2 SIRNA Assays ›
Gene Symbol:
Zfp36l2 LOC100911319
Gene Name:
zinc finger protein 36, C3H type-like 2
zinc finger protein 36, C3H1 type-like 2-like
Gene Aliases:
-
-
Chromosome Location:
Chr.6: 7417416 - 7423484 on Build Rnor_6.0
Chromosome Location:
Chr.7: 2350068 - 2354108 on Build Rnor_6.0
UniGene ID:
Rn.22055
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS341726
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp36l2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001036626.1 NP_001031703.1 1 262
XM_008764455.1 XP_008762677.1 1 2290
GenBank mRNA CV112816.1 - 1 262
Gene Symbol: LOC100911319
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750261.1 XP_003750309.1 1 262
XM_008765049.1 XP_008763271.1 1 255

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
mRNA catabolic process
hemopoiesis
T cell differentiation in thymus
somatic stem cell population maintenance
regulation of mRNA stability
somatic stem cell division
definitive hemopoiesis
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
negative regulation of stem cell differentiation
AU-rich element binding
mRNA 3'-UTR AU-rich region binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0