This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100361056 SIRNA Assays ›
Gene Symbol:
LOC100361056 Tmem60
Gene Name:
transmembrane protein 60-like
transmembrane protein 60
Gene Aliases:
-
RGD1308939
Chromosome Location:
Chr.16: 20931522 - 20932708 on Build Rnor_6.0
Chromosome Location:
Chr.4: 10818024 - 10822879 on Build Rnor_6.0
UniGene ID:
-
Rn.9415
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS341634
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100361056
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252979.2 XP_006253041.1 1 654
Gene Symbol: Tmem60
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191610.1 NP_001178539.1 2 332

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0