This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FCGR2B SIRNA Assays ›
Gene Symbol:
Fcgr2b
Gene Name:
Fc fragment of IgG, low affinity IIb, receptor
Gene Aliases:
CD32, FCRII, FcgammaRIIB2, Fcgr2
Chromosome Location:
Chr.13: 89329298 - 89344169 on Build Rnor_6.0
UniGene ID:
Rn.33323
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS341090
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fcgr2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175756.1 NP_786932.1 3 370
XM_006250233.1 XP_006250295.1 3 387
XM_006250234.2 XP_006250296.1 4 400
XM_006250235.1 XP_006250297.1 3 384
GenBank mRNA BC091113.1 - 3 364 7323350
EF424788.1 - 3 339
FQ228752.1 - 3 389
X73371.1 - 3 404

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of type I hypersensitivity
immune complex clearance
negative regulation of immunoglobulin production
negative regulation of inflammatory response to antigenic stimulus
negative regulation of acute inflammatory response to antigenic stimulus
negative regulation of humoral immune response mediated by circulating immunoglobulin
receptor-mediated endocytosis
phagocytosis, engulfment
defense response
immune response
cell surface receptor signaling pathway
response to bacterium
immunoglobulin mediated immune response
antigen processing and presentation of exogenous peptide antigen via MHC class II
negative regulation of B cell proliferation
response to estradiol
endocytic recycling
negative regulation of interleukin-10 production
negative regulation of neutrophil apoptotic process
mast cell activation
negative regulation of phagocytosis
positive regulation of phagocytosis
negative regulation of immune response
cellular response to molecule of bacterial origin
cellular response to lipopolysaccharide
IgG binding
immunoglobulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0