This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APTX SIRNA Assays ›
Gene Symbol:
Aptx
Gene Name:
aprataxin
Gene Aliases:
-
Chromosome Location:
Chr.5: 56987714 - 57009442 on Build Rnor_6.0
UniGene ID:
Rn.50459
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS340874
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aptx
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148889.2 NP_683687.1 7 1051
XM_006238026.2 XP_006238088.1 7 1407
XM_006238027.2 XP_006238089.1 6 1443
XM_006238028.2 XP_006238090.1 6 1290
XM_006238029.2 XP_006238091.1 5 624
XM_008763607.1 XP_008761829.1 5 1302
GenBank mRNA AF398235.1 - 6 797
BC078716.1 - 7 1051 7132044
FQ214915.1 - 3 424

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

single strand break repair
DNA catabolic process, exonucleolytic
DNA ligation
double-strand break repair
cellular response to DNA damage stimulus
biological_process
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
polynucleotide 3' dephosphorylation
molecular_function
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
polynucleotide 3'-phosphatase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0