This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IFNG SIRNA Assays ›
Gene Symbol:
Ifng
Gene Name:
interferon gamma
Gene Aliases:
IFNG2
Chromosome Location:
Chr.7: 61337383 - 61341419 on Build Rnor_6.0
UniGene ID:
Rn.10795
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS340871
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ifng
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138880.2 NP_620235.1 3 300
GenBank mRNA AF010466.1 - 3 300

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus, translocation
negative regulation of transcription from RNA polymerase II promoter
neutrophil apoptotic process
regulation of the force of heart contraction
adaptive immune response
CD8-positive, alpha-beta T cell differentiation involved in immune response
regulation of transcription, DNA-templated
apoptotic process
inflammatory cell apoptotic process
humoral immune response
cell cycle arrest
positive regulation of cell proliferation
negative regulation of cell proliferation
response to virus
antigen processing and presentation
neutrophil chemotaxis
negative regulation of epithelial cell differentiation
endoplasmic reticulum unfolded protein response
negative regulation of myelination
positive regulation of synaptic transmission, cholinergic
negative regulation of interleukin-17 production
positive regulation of interleukin-12 production
positive regulation of interleukin-23 production
positive regulation of tumor necrosis factor production
positive regulation of peptidyl-serine phosphorylation of STAT protein
positive regulation of smooth muscle cell apoptotic process
positive regulation of T cell proliferation
response to drug
positive regulation of tyrosine phosphorylation of Stat1 protein
defense response to bacterium
defense response to protozoan
negative regulation of growth of symbiont in host
negative regulation of growth of symbiont involved in interaction with host
positive regulation of chemokine biosynthetic process
positive regulation of interleukin-12 biosynthetic process
positive regulation of MHC class II biosynthetic process
positive regulation of interleukin-6 biosynthetic process
positive regulation of nitric oxide biosynthetic process
positive regulation of neuron differentiation
positive regulation of osteoclast differentiation
positive regulation of cell adhesion
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of isotype switching to IgG isotypes
negative regulation of smooth muscle cell proliferation
positive regulation of interleukin-1 beta secretion
regulation of immune response
regulation of insulin secretion
sensory perception of mechanical stimulus
positive regulation of membrane protein ectodomain proteolysis
defense response to virus
positive regulation of killing of cells of other organism
positive regulation of fructose 1,6-bisphosphate 1-phosphatase activity
positive regulation of fructose 1,6-bisphosphate metabolic process
positive regulation of vitamin D biosynthetic process
positive regulation of calcidiol 1-monooxygenase activity
cellular response to lipopolysaccharide
cellular response to interleukin-18
extrinsic apoptotic signaling pathway
regulation of neuronal action potential
positive regulation of tumor necrosis factor (ligand) superfamily member 11 production
regulation of hepatocyte proliferation
cytokine activity
interferon-gamma receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0