This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APOA2 SIRNA Assays ›
Gene Symbol:
Apoa2
Gene Name:
apolipoprotein A-II
Gene Aliases:
APOAII, Apo-AII, ApoA-II
Chromosome Location:
Chr.13: 89596872 - 89598805 on Build Rnor_6.0
UniGene ID:
Rn.89304
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS340855
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Apoa2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013112.1 NP_037244.1 2 127
XM_006250238.2 XP_006250300.1 3 261
XM_006250239.2 XP_006250301.1 3 267
GenBank mRNA FQ209753.1 - 3 192
FQ209821.1 - 3 192
FQ210214.1 - 3 185
FQ210295.1 - 3 189
FQ210325.1 - 3 212
FQ210458.1 - 3 189
FQ210462.1 - 3 190
FQ210537.1 - 2 186
FQ210542.1 - 2 315
FQ210648.1 - 3 189
FQ210885.1 - 3 219
FQ211006.1 - 3 186
FQ211265.1 - 3 189
FQ211383.1 - 3 191
FQ218158.1 - 3 189
FQ218273.1 - 3 192
FQ218326.1 - 3 186
FQ218756.1 - 3 193
FQ221741.1 - 3 192
FQ223965.1 - 3 186
M28615.1 - 2 96
X03468.1 - 2 127

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

acute inflammatory response
negative regulation of cytokine secretion involved in immune response
protein folding
phosphatidylcholine biosynthetic process
lipid transport
cholesterol metabolic process
phospholipid catabolic process
response to glucose
positive regulation of cholesterol esterification
negative regulation of very-low-density lipoprotein particle remodeling
protein oxidation
peptidyl-methionine modification
regulation of intestinal cholesterol absorption
cholesterol transport
organ regeneration
regulation of protein stability
negative regulation of cholesterol transport
cholesterol efflux
phospholipid efflux
triglyceride-rich lipoprotein particle remodeling
low-density lipoprotein particle remodeling
high-density lipoprotein particle remodeling
high-density lipoprotein particle assembly
high-density lipoprotein particle clearance
lipoprotein metabolic process
response to drug
cholesterol homeostasis
positive regulation of catalytic activity
response to estrogen
reverse cholesterol transport
positive regulation of interleukin-8 biosynthetic process
diacylglycerol catabolic process
negative regulation of lipid catabolic process
positive regulation of lipid catabolic process
response to glucocorticoid
negative regulation of lipase activity
negative regulation of cholesterol import
negative regulation of cholesterol transporter activity
lipid transporter activity
phospholipid binding
high-density lipoprotein particle binding
lipid binding
cholesterol binding
cholesterol transporter activity
phosphatidylcholine binding
apolipoprotein receptor binding
protein homodimerization activity
protein heterodimerization activity
lipase inhibitor activity
phosphatidylcholine-sterol O-acyltransferase activator activity
high-density lipoprotein particle receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0