This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHRNB4 SIRNA Assays ›
Gene Symbol:
Chrnb4
Gene Name:
cholinergic receptor, nicotinic, beta 4 (neuronal)
Gene Aliases:
-
Chromosome Location:
Chr.8: 59610489 - 59629073 on Build Rnor_6.0
UniGene ID:
Rn.9695
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS340764
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chrnb4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052806.2 NP_434693.1 3, 4 246
GenBank mRNA AY574260.1 - 3, 4 246
U42976.1 - 306
X15834.1 - 358

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_148944, AF492840, BC138851, AY574269, AK140071.

Gene Ontology Categories:

Function(s) Process(es)

action potential
cation transport
smooth muscle contraction
regulation of smooth muscle contraction
signal transduction
locomotory behavior
response to nicotine
behavioral response to nicotine
regulation of membrane potential
protein heterooligomerization
synaptic transmission involved in micturition
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
drug binding
acetylcholine receptor activity
acetylcholine binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm