siRNA Details
Gene Symbol: Ghsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032075.3 | NP_114464.1 | 2 | 1076 | ||
GenBank mRNA | AB001982.1 | - | 2 | 1076 | ||
U94321.1 | - | 1 | 3110 |

Gene Symbol: Ghsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032075.3 | NP_114464.1 | 2 | 1076 | ||
GenBank mRNA | AB001982.1 | - | 2 | 1076 | ||
U94321.1 | - | 1 | 3110 |