This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DECR2 SIRNA Assays ›
Gene Symbol:
Decr2
Gene Name:
2,4-dienoyl CoA reductase 2, peroxisomal
Gene Aliases:
Dcrakl, putativeperoxisomal24-dienoyl-CoAreductase
Chromosome Location:
Chr.10: 15451850 - 15460215 on Build Rnor_6.0
UniGene ID:
Rn.144598
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS340019
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Decr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_171996.3 NP_741993.1 5 493
XM_006246024.2 XP_006246086.1 6 693
GenBank mRNA AF044574.1 - 5 415
BC070959.1 - 5 494 7097883
FQ218511.1 - 5 468

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

unsaturated fatty acid biosynthetic process
oxidation-reduction process
receptor binding
2,4-dienoyl-CoA reductase (NADPH) activity
trans-2-enoyl-CoA reductase (NADPH) activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header