This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NR1H3 SIRNA Assays ›
Gene Symbol:
Nr1h3
Gene Name:
nuclear receptor subfamily 1, group H, member 3
Gene Aliases:
LXRalpha
Chromosome Location:
Chr.3: 80004130 - 80014197 on Build Rnor_6.0
UniGene ID:
Rn.11209
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS339827
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nr1h3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031627.2 NP_113815.2 9 1223
XM_006234533.2 XP_006234595.1 10 1401
XM_006234534.1 XP_006234596.1 10 1342
XM_006234535.2 XP_006234597.1 10 1543
XM_006234537.1 XP_006234599.1 9 1013
XM_008762013.1 XP_008760235.1 9 1133
GenBank mRNA BC088750.1 - 9 1223 7314121
U11685.1 - 9 1230

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001251935, NM_001130101, NM_005693, NM_001130102, NM_001251934, and others.
This siRNA also targets the following transcripts in Mouse: AJ132601.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
lipid metabolic process
insulin receptor signaling pathway
positive regulation of triglyceride biosynthetic process
positive regulation of receptor biosynthetic process
positive regulation of cholesterol efflux
negative regulation of cholesterol storage
intracellular receptor signaling pathway
positive regulation of cellular protein metabolic process
negative regulation of lipid transport
positive regulation of cholesterol transport
response to progesterone
positive regulation of toll-like receptor 4 signaling pathway
cholesterol homeostasis
negative regulation of macrophage activation
apoptotic cell clearance
steroid hormone mediated signaling pathway
cellular lipid metabolic process
positive regulation of fatty acid biosynthetic process
negative regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of pinocytosis
negative regulation of inflammatory response
positive regulation of lipoprotein lipase activity
lipid homeostasis
sterol homeostasis
triglyceride homeostasis
cellular response to lipopolysaccharide
negative regulation of pancreatic juice secretion
negative regulation of secretion of lysosomal enzymes
regulation of cholesterol homeostasis
positive regulation of cholesterol homeostasis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
zinc ion binding
sterol response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding
retinoid X receptor binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0