This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ESPN SIRNA Assays ›
Gene Symbol:
Espn
Gene Name:
espin
Gene Aliases:
Je
Chromosome Location:
Chr.5: 169293356 - 169331338 on Build Rnor_6.0
UniGene ID:
Rn.91373
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS339794
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Espn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239476.2 XP_006239538.1 7 1396
XM_006239479.2 XP_006239541.1 7 1396
XM_008764375.1 XP_008762597.1 7 1405
XM_008764376.1 XP_008762598.1 7 1405
XM_008764377.1 XP_008762599.1 7 1405
XM_008764378.1 XP_008762600.1 7 1405
XM_008764379.1 XP_008762601.1 7 1405
XM_008764380.1 XP_008762602.1 7 1405
GenBank mRNA AF540946.1 - 2 225
AF540947.1 - 2 225
AF540948.1 - 2 225
AF540949.1 - 2 225
U46007.1 - 7 1219

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
sensory perception of sound
locomotory behavior
parallel actin filament bundle assembly
actin filament bundle assembly
positive regulation of filopodium assembly
negative regulation of cytoskeleton organization
SH3 domain binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0