This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ESPN SIRNA Assays ›
Gene Symbol:
Espn
Gene Name:
espin
Gene Aliases:
Je
Chromosome Location:
Chr.5: 169293356 - 169331338 on Build Rnor_6.0
UniGene ID:
Rn.91373
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS339792
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Espn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239476.2 XP_006239538.1 1 340
XM_006239479.2 XP_006239541.1 1 340
XM_008764375.1 XP_008762597.1 1 340
XM_008764376.1 XP_008762598.1 1 340
XM_008764377.1 XP_008762599.1 1 340
XM_008764378.1 XP_008762600.1 1 340
XM_008764379.1 XP_008762601.1 1 340
XM_008764380.1 XP_008762602.1 1 340
GenBank mRNA U46007.1 - 1 166

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
sensory perception of sound
locomotory behavior
parallel actin filament bundle assembly
actin filament bundle assembly
positive regulation of filopodium assembly
negative regulation of cytoskeleton organization
SH3 domain binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0