This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CACNA1E SIRNA Assays ›
Gene Symbol:
Cacna1e
Gene Name:
calcium channel, voltage-dependent, R type, alpha 1E subunit
Gene Aliases:
CACHA1E
Chromosome Location:
Chr.13: 71899445 - 72485108 on Build Rnor_6.0
UniGene ID:
Rn.10742
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS339742
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cacna1e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019294.2 NP_062167.2 27 3929
XM_006250069.2 XP_006250131.1 30 5303
GenBank mRNA AY029412.1 - 27 3929
L15453.1 - 28 4026

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

behavioral fear response
regulation of heart rate
calcium ion transport
chemical synaptic transmission
locomotory behavior
visual learning
transmission of nerve impulse
sensory perception of pain
sperm motility
regulation of ion transmembrane transport
glucose homeostasis
fear response
response to pain
behavioral response to pain
neurological system process
regulation of insulin secretion involved in cellular response to glucose stimulus
calcium ion import
calcium ion transmembrane transport
membrane depolarization during action potential
regulation of somatostatin secretion
voltage-gated calcium channel activity
calcium ion binding
high voltage-gated calcium channel activity
low voltage-gated calcium channel activity
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0