This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CRLS1 SIRNA Assays ›
Gene Symbol:
Crls1
Gene Name:
cardiolipin synthase 1
Gene Aliases:
RGD1311037
Chromosome Location:
Chr.3: 125503638 - 125522446 on Build Rnor_6.0
UniGene ID:
Rn.35982
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS339026
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Crls1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014258.1 NP_001014280.1 3 675
XM_006235078.2 XP_006235140.1 3 869
XM_006235079.2 XP_006235141.1 3 858
XM_006235080.2 XP_006235142.1 3 358
GenBank mRNA BC085849.1 - 3 675 7189638

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phospholipid biosynthetic process
phosphotransferase activity, for other substituted phosphate groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0