This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SFXN4 SIRNA Assays ›
Gene Symbol:
Sfxn4
Gene Name:
sideroflexin 4
Gene Aliases:
-
Chromosome Location:
Chr.1: 282213610 - 282236854 on Build Rnor_6.0
UniGene ID:
Rn.13517
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS337989
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sfxn4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108527.1 NP_001101997.1 7 418
XM_006231684.2 XP_006231746.1 6 2479
XM_006231685.2 XP_006231747.1 6 1315
XM_008760590.1 XP_008758812.1 5 2213
GenBank mRNA FQ215895.1 - 7 407

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_053198, AF325263, AK148530, BC089535.

Gene Ontology Categories:

Function(s) Process(es)

ion transmembrane transport
ion transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0