This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MOB3C SIRNA Assays ›
Gene Symbol:
Mob3c
Gene Name:
MOB kinase activator 3C
Gene Aliases:
Mobkl2c
Chromosome Location:
Chr.5: 134679858 - 134687896 on Build Rnor_6.0
UniGene ID:
Rn.162087
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS337087
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mob3c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107960.1 NP_001101430.1 2 284
XM_006238657.2 XP_006238719.1 1 1200
XM_006238658.2 XP_006238720.1 2 705
XM_006238659.2 XP_006238721.1 2 2202
XM_008763977.1 XP_008762199.1 1 1200

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175308, BC099740, BC137587, AK148476, BC137589, and others.

Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0