This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CFAP20 SIRNA Assays ›
Gene Symbol:
Cfap20
Gene Name:
cilia and flagella associated protein 20
Gene Aliases:
Gtl3
Chromosome Location:
Chr.19: 10024938 - 10038838 on Build Rnor_6.0
UniGene ID:
Rn.6708
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS336425
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cfap20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037978.1 NP_001033067.1 2, 3 342
XM_006255083.1 XP_006255145.1 2, 3 428
GenBank mRNA BC099768.1 - 2, 3 342 7932140

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyglutamylation
cilium morphogenesis
regulation of cilium beat frequency involved in ciliary motility
positive regulation of cell motility
positive regulation of feeding behavior
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0