This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TBX5 SIRNA Assays ›
Gene Symbol:
Tbx5
Gene Name:
T-box 5
Gene Aliases:
-
Chromosome Location:
Chr.12: 42059688 - 42148226 on Build Rnor_6.0
UniGene ID:
Rn.6327
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS336083
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tbx5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009964.1 NP_001009964.1 8 1119
XM_006249387.2 XP_006249449.1 9 1614
XM_006249388.2 XP_006249450.1 9 1743
XM_006249389.2 XP_006249451.1 10 1918
XM_006249390.2 XP_006249452.1 10 1789
XM_006249391.2 XP_006249453.1 9 1763
XM_006249392.2 XP_006249454.1 9 1308
XM_008769230.1 XP_008767452.1 10 1766
XM_008769231.1 XP_008767453.1 9 1419
GenBank mRNA AY859491.1 - 8 1119

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

morphogenesis of an epithelium
bundle of His development
atrioventricular valve morphogenesis
endocardial cushion development
cardiac left ventricle formation
ventricular cardiac muscle tissue development
ventricular septum development
atrial septum development
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
cell-cell signaling
pattern specification process
heart development
negative regulation of cell proliferation
apoptotic nuclear changes
lung development
embryonic limb morphogenesis
negative regulation of cell migration
embryonic forelimb morphogenesis
forelimb morphogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of cardioblast differentiation
cardiac muscle cell differentiation
pericardium development
negative regulation of cardiac muscle cell proliferation
positive regulation of cardiac muscle cell proliferation
transdifferentiation
atrial septum morphogenesis
positive regulation of secondary heart field cardioblast proliferation
regulation of execution phase of apoptosis
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0