This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNAB2 SIRNA Assays ›
Gene Symbol:
Kcnab2
Gene Name:
potassium channel, voltage-gated shaker-related subfamily A regulatory beta subunit 2
Gene Aliases:
-
Chromosome Location:
Chr.5: 169570327 - 169658875 on Build Rnor_6.0
UniGene ID:
Rn.10757
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS334931
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnab2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017304.2 NP_059000.1 14 1343
XM_008764342.1 XP_008762564.1 14 1109
XM_008764343.1 XP_008762565.1 14 1362
XM_008764344.1 XP_008762566.1 13 1066
XM_008764345.1 XP_008762567.1 12 951
GenBank mRNA X76724.1 - 14 1453

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
neuromuscular process
protein heterooligomerization
oxidation-reduction process
NADPH oxidation
potassium ion transmembrane transport
regulation of potassium ion transmembrane transport
regulation of protein localization to cell surface
aldo-keto reductase (NADP) activity
voltage-gated potassium channel activity
protein binding
potassium channel regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0