This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PACSIN1 SIRNA Assays ›
Gene Symbol:
Pacsin1
Gene Name:
protein kinase C and casein kinase substrate in neurons 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 7241248 - 7286702 on Build Rnor_6.0
UniGene ID:
Rn.161729
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS334894
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pacsin1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017294.1 NP_058990.1 5 714
GenBank mRNA AF104402.1 - 5 714

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
actin filament organization
regulation of endocytosis
negative regulation of endocytosis
synaptic vesicle endocytosis
neuron development
neuron projection morphogenesis
protein localization to membrane
establishment of protein localization to plasma membrane
membrane tubulation
positive regulation of dendrite development
protein binding
phospholipid binding
cytoskeletal protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0