This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C1QB SIRNA Assays ›
Gene Symbol:
C1qb
Gene Name:
complement component 1, q subcomponent, B chain
Gene Aliases:
-
Chromosome Location:
Chr.5: 155246444 - 155251995 on Build Rnor_6.0
UniGene ID:
Rn.6702
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS334874
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C1qb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019262.1 NP_062135.1 3 676
GenBank mRNA FQ227589.1 - 3 601
FQ228182.1 - 3 536
FQ234810.1 - 3 615
X71127.1 - 3 676

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune response
complement activation, classical pathway
brain development
aging
response to organic substance
innate immune response
inner ear development
response to glucocorticoid
molecular_function
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0