This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCN1B SIRNA Assays ›
Gene Symbol:
Scn1b
Gene Name:
sodium channel, voltage-gated, type I, beta subunit
Gene Aliases:
-
Chromosome Location:
Chr.1: 89550738 - 89560469 on Build Rnor_6.0
UniGene ID:
Rn.4958
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS334871
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Scn1b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271045.1 NP_001257974.1 4, 5 723
NM_001271046.2 NP_001257975.1 4, 5 590
NM_017288.2 NP_058984.1 4, 5 804
GenBank mRNA BC094523.1 - 4, 5 723 7316686
FQ215379.1 - 2, 3 381
FQ216374.1 - 4, 5 590
M91808.1 - 4, 5 803

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of sodium ion transport
axon guidance
positive regulation of sodium ion transport
positive regulation of neuron projection development
neuronal action potential propagation
corticospinal neuron axon guidance
sodium ion transmembrane transport
locomotion
response to pyrethroid
membrane depolarization
cardiac muscle contraction
regulation of ventricular cardiac muscle cell membrane repolarization
regulation of atrial cardiac muscle cell membrane depolarization
cardiac conduction
cardiac muscle cell action potential involved in contraction
membrane depolarization during cardiac muscle cell action potential
membrane depolarization during Purkinje myocyte cell action potential
regulation of heart rate by cardiac conduction
regulation of sodium ion transmembrane transporter activity
voltage-gated sodium channel activity
protein binding
sodium channel regulator activity
sodium channel inhibitor activity
voltage-gated sodium channel activity involved in cardiac muscle cell action potential
voltage-gated sodium channel activity involved in Purkinje myocyte action potential

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0