This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NFYA SIRNA Assays ›
Gene Symbol:
Nfya
Gene Name:
nuclear transcription factor-Y alpha
Gene Aliases:
CBF-A
Chromosome Location:
Chr.9: 14551752 - 14577002 on Build Rnor_6.0
UniGene ID:
Rn.10747
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS334499
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nfya
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012865.1 NP_036997.1 7 765
XM_006244409.2 XP_006244471.1 7 790
XM_006244410.1 XP_006244472.1 7 786
XM_006244411.2 XP_006244473.1 7 780
XM_006244412.2 XP_006244474.1 7 777
XM_006244413.1 XP_006244475.1 6 698
XM_008766839.1 XP_008765061.1 6 686
GenBank mRNA FQ210330.1 - 6 703
FQ223023.1 - 7 722
M34238.1 - 7 765

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010913, NM_001110832, AK143723, BC029695, M86214, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
regulation of stem cell population maintenance
positive regulation of stem cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription cofactor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
transcription regulatory region DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0