This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IRF8 SIRNA Assays ›
Gene Symbol:
Irf8
Gene Name:
interferon regulatory factor 8
Gene Aliases:
ICSBP, Icsbp1
Chromosome Location:
Chr.19: 54314860 - 54336643 on Build Rnor_6.0
UniGene ID:
Rn.228738
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS333775
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Irf8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008722.1 NP_001008722.1 7, 8 1086
XM_006255737.2 XP_006255799.1 8, 9 1134
GenBank mRNA AB120371.1 - 7, 8 1086

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008320, AK152906, AK153166, AK159835, AK157147, and others.

Gene Ontology Categories:

Function(s) Process(es)

phagocytosis
immune response
response to bacterium
myeloid cell differentiation
positive regulation of interferon-gamma production
positive regulation of interleukin-12 production
defense response to bacterium
defense response to protozoan
negative regulation of growth of symbiont in host
positive regulation of transcription, DNA-templated
positive regulation of transcription initiation from RNA polymerase II promoter
cellular response to lipopolysaccharide
regulatory region DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0