This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VTN SIRNA Assays ›
Gene Symbol:
Vtn
Gene Name:
vitronectin
Gene Aliases:
Aa1018, Vn
Chromosome Location:
Chr.10: 65767960 - 65771040 on Build Rnor_6.0
UniGene ID:
Rn.87493
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS333693
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vtn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019156.2 NP_062029.2 7 1281
GenBank mRNA AY318963.1 - 1278
BC105821.1 - 7 1281 7371104
FQ209609.1 - 7 1294
FQ209682.1 - 7 1289
FQ209762.1 - 3 455
FQ210146.1 - 5 901
FQ210518.1 - 7 1286
FQ211325.1 - 3 415
FQ212575.1 - 7 1281
FQ218318.1 - 7 1293
FQ218401.1 - 5 853
FQ219297.1 - 7 1296
U44845.1 - 7 1271

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
immune response
cell-matrix adhesion
positive regulation of cell-substrate adhesion
negative regulation of endopeptidase activity
positive regulation of smooth muscle cell migration
extracellular matrix organization
positive regulation of protein binding
cell adhesion mediated by integrin
endodermal cell differentiation
positive regulation of receptor-mediated endocytosis
positive regulation of peptidyl-tyrosine phosphorylation
smooth muscle cell-matrix adhesion
scavenger receptor activity
integrin binding
polysaccharide binding
extracellular matrix binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0