siRNA Details
Gene Symbol: Aldh7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271105.1 | NP_001258034.1 | 15 | 1411 |

More Information
Other Species Targeted

Gene Ontology Categories:

Gene Symbol: Aldh7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271105.1 | NP_001258034.1 | 15 | 1411 |