This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ACTR2 SIRNA Assays ›
Gene Symbol:
Actr2
Gene Name:
ARP2 actin-related protein 2 homolog (yeast)
Gene Aliases:
-
Chromosome Location:
Chr.14: 104340783 - 104375648 on Build Rnor_6.0
UniGene ID:
Rn.102249
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS333428
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Actr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009268.1 NP_001009268.1 3 379
GenBank mRNA BC088442.1 - 3 379 7126325
FQ212814.1 - 4 343
FQ219071.1 - 3 328
FQ219838.1 - 3 345
FQ225424.1 - 3 350
FQ226083.1 - 3 325
FQ226422.1 - 3 328
FQ227488.1 - 3 377
FQ227830.1 - 3 328
FQ230874.1 - 3 363
FQ231828.1 - 3 330
FQ231874.1 - 3 363
FQ234230.1 - 3 330
FQ235071.1 - 3 350
FQ235201.1 - 3 345

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

establishment or maintenance of cell polarity
asymmetric cell division
meiotic chromosome movement towards spindle pole
cytosolic transport
actin cytoskeleton organization
meiotic cytokinesis
Arp2/3 complex-mediated actin nucleation
cilium assembly
meiotic cell cycle
spindle localization
actin binding
structural constituent of cytoskeleton
ATP binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0