This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FMNL1 SIRNA Assays ›
Gene Symbol:
Fmnl1
Gene Name:
formin-like 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 91253876 - 91281422 on Build Rnor_6.0
UniGene ID:
Rn.45477
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS333078
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fmnl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105846.1 NP_001099316.1 1, 2 274
XM_006247498.2 XP_006247560.1 1, 2 485
XM_006247499.2 XP_006247561.1 1, 2 486
XM_006247500.2 XP_006247562.1 1, 2 485
XM_006247501.2 XP_006247563.1 1, 2 485
XM_006247502.2 XP_006247564.1 1, 2 485
XM_008768266.1 XP_008766488.1 1, 2 484
XM_008768267.1 XP_008766489.1 1, 2 485

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

substrate-dependent cell migration
regulation of cell shape
cellular process
cortical actin cytoskeleton organization
actin filament severing
profilin binding
GTPase activating protein binding
Rac GTPase binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0