This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ELAVL3 SIRNA Assays ›
Gene Symbol:
Elavl3
Gene Name:
ELAV like neuron-specific RNA binding protein 3
Gene Aliases:
Huc
Chromosome Location:
Chr.8: 23029507 - 23063359 on Build Rnor_6.0
UniGene ID:
Rn.22586
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS332866
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Elavl3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172324.1 NP_758827.1 3 317
XM_006242616.2 XP_006242678.1 3 661
XM_008765920.1 XP_008764142.1 3 504
GenBank mRNA AB097198.1 - 3 317

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001420, NM_032281, AY034002, BC014144, AY036909, and others.
This siRNA also targets the following transcripts in Mouse: NM_010487, U29148, BC052097, U29149, AK090281.

Gene Ontology Categories:

Process(es)

nucleotide binding
AU-rich element binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0