This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MIOX SIRNA Assays ›
Gene Symbol:
Miox
Gene Name:
myo-inositol oxygenase
Gene Aliases:
Aldrl6, RSOR
Chromosome Location:
Chr.7: 130308707 - 130311214 on Build Rnor_6.0
UniGene ID:
Rn.19959
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS332620
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Miox
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145771.2 NP_665714.2 3 124
XM_008765674.1 XP_008763896.1 3 308
GenBank mRNA AF197128.1 - 3 118
AY738259.1 - 2 118
BC078840.1 - 3 124 7110224
FQ219187.1 - 2 113

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

inositol catabolic process
oxidation-reduction process
aldo-keto reductase (NADP) activity
ferric iron binding
oxidoreductase activity, acting on NAD(P)H
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen
inositol oxygenase activity
NADP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0