This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRKAR2B SIRNA Assays ›
Gene Symbol:
Prkar2b
Gene Name:
protein kinase, cAMP dependent regulatory, type II beta
Gene Aliases:
RATDNA
Chromosome Location:
Chr.6: 51265509 - 51356383 on Build Rnor_6.0
UniGene ID:
Rn.164375
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS332448
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prkar2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030020.1 NP_001025191.1 3, 4 385
GenBank mRNA BC099223.1 - 2, 3 128 7388087
M12492.1 - 3, 4 379

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
learning
regulation of protein kinase activity
response to antipsychotic drug
response to clozapine
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
protein domain specific binding
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0