This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100911685 SIRNA Assays ›
Gene Symbol:
LOC100911685 Gspt1
Gene Name:
eukaryotic peptide chain release factor GTP-binding subunit ERF3B-like
G1 to S phase transition 1
Gene Aliases:
-
-
Chromosome Location:
Chr.10: 4411146 - 4446198 on Build Rnor_6.0
Chromosome Location:
Chr.10: 4313100 - 4347661 on Build Rnor_6.0
UniGene ID:
-
Rn.72491
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID RSS332356
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100911685
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750763.3 XP_003750811.1 1600
XM_006245740.2 XP_006245802.1 1603
Gene Symbol: Gspt1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003978.1 NP_001003978.1 8 1177
GenBank mRNA BC079092.1 - 8 1177 7123818

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001130006, NM_002094, NM_018094, NM_001130007, AK315267, and others.
This siRNA also targets the following transcripts in Mouse: NM_001130008, NM_146066, BC031640, BC028325, AK148603, and others.

Gene Ontology Categories:

Function(s) Process(es)

translational termination
protein methylation
metabolic process
translation release factor activity
poly(A) RNA binding
GTPase activity
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0