This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CKM SIRNA Assays ›
Gene Symbol:
Ckm
Gene Name:
creatine kinase, muscle
Gene Aliases:
Ckmm
Chromosome Location:
Chr.1: 80321585 - 80331841 on Build Rnor_6.0
UniGene ID:
Rn.10756
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS332318
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ckm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012530.2 NP_036662.1 2 111
GenBank mRNA BC062058.1 - 2 62 5598226
DN931989.1 - 2 110
FQ214594.1 - 2 87
FQ214655.1 - 2 87
FQ214750.1 - 2 87
FQ214931.1 - 2 88
FQ215410.1 - 2 87
FQ216507.1 - 2 82
FQ224918.1 - 2 88
M10140.1 - 2 84

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007710, X03233, BC132426, AK137460, AK009950, and others.

Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
phosphocreatine biosynthetic process
creatine kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0