This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADAP1 SIRNA Assays ›
Gene Symbol:
Adap1
Gene Name:
ArfGAP with dual PH domains 1
Gene Aliases:
Centa1, p42IP4
Chromosome Location:
Chr.12: 17416327 - 17468212 on Build Rnor_6.0
UniGene ID:
Rn.10539
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS332159
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 491.00
Your Price
Online offer:
491.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adap1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133567.2 NP_598251.1 5 566
GenBank mRNA AF123047.1 - 5 493
AJ007422.1 - 5 493
AJ007616.1 - 5 493
BC099775.1 - 5 566 7933317

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of GTPase activity
positive regulation of GTPase activity
inositol phosphate-mediated signaling
phosphatidylinositol-3,4,5-trisphosphate binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0