This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD79B SIRNA Assays ›
Gene Symbol:
Cd79b
Gene Name:
Cd79b molecule, immunoglobulin-associated beta
Gene Aliases:
B29, Igb
Chromosome Location:
Chr.10: 94497445 - 94500591 on Build Rnor_6.0
UniGene ID:
Rn.62607
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS332144
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd79b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133533.2 NP_598217.1 3 439
GenBank mRNA BC088115.1 - 3 439 7325406
FQ226266.1 - 3 436
FQ227061.1 - 3 434
FQ227788.1 - 3 428
FQ230261.1 - 3 430
FQ232054.1 - 3 430
FQ232680.1 - 3 430
FQ232790.1 - 3 453
FQ233507.1 - 3 428
FQ234082.1 - 3 413
FQ234487.1 - 3 428

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

B cell receptor signaling pathway
transmembrane signaling receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header