This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHRNA2 SIRNA Assays ›
Gene Symbol:
Chrna2
Gene Name:
cholinergic receptor, nicotinic, alpha 2 (neuronal)
Gene Aliases:
ACHR
Chromosome Location:
Chr.15: 42808897 - 42825857 on Build Rnor_6.0
UniGene ID:
Rn.9713
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS332132
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chrna2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133420.1 NP_596911.1 6 720
XM_008770735.1 XP_008768957.1 6 562
GenBank mRNA AY574253.1 - 5 416
L10077.1 - 6 720

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cation transport
signal transduction
chemical synaptic transmission
protein heterooligomerization
cation transmembrane transport
acetylcholine-activated cation-selective channel activity
drug binding
acetylcholine receptor activity
acetylcholine binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header