This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CNGA1 SIRNA Assays ›
Gene Symbol:
Cnga1
Gene Name:
cyclic nucleotide gated channel alpha 1
Gene Aliases:
Cncg, HCN
Chromosome Location:
Chr.14: 38155771 - 38171107 on Build Rnor_6.0
UniGene ID:
Rn.54534
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS331625
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnga1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053497.1 NP_445949.1 8 985
GenBank mRNA U93851.1 - 8 985

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
visual perception
phototransduction, visible light
ion transmembrane transport
regulation of cytosolic calcium ion concentration
membrane depolarization
potassium ion transmembrane transport
ion channel activity
intracellular cyclic nucleotide activated cation channel activity
intracellular cAMP activated cation channel activity
intracellular cGMP activated cation channel activity
voltage-gated potassium channel activity
cGMP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0