This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHN1 SIRNA Assays ›
Gene Symbol:
Chn1
Gene Name:
chimerin 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 60512360 - 60668281 on Build Rnor_6.0
UniGene ID:
Rn.11166
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS331337
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chn1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032083.1 NP_114472.1 3 772
XM_006234386.2 XP_006234448.1 9 1488
XM_006234388.2 XP_006234450.1 9 728
XM_006234389.2 XP_006234451.1 9 1431
XM_008761932.1 XP_008760154.1 6 1488
GenBank mRNA BC094519.1 - 3 821 7315609
X67250.1 - 3 772

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
motor neuron axon guidance
intracellular signal transduction
positive regulation of GTPase activity
ephrin receptor signaling pathway
regulation of axonogenesis
phorbol ester receptor activity
GTPase activator activity
metal ion binding
ephrin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0